Dimethyl malonate, 99+%
Dimethyl malonate, 99+%
Dimethyl malonate, 99+%
Thermo Scientific Chemicals

Dimethyl malonate, 99+%

CAS: 108-59-8 | C5H8O4 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 220702500
Price (USD)
52.40
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.40
Each
Chemical Identifiers
CAS108-59-8
IUPAC Name1,3-dimethyl propanedioate
Molecular FormulaC5H8O4
InChI KeyBEPAFCGSDWSTEL-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.1 % (K.F.)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Free acid=<0.1 % (as malonic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only