1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
Thermo Scientific Chemicals

1,2-Propanediol, 99+%, for analysis

CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
產品號碼 220870250
價格 (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
檢視更多
規格Specification Sheet規格表
Water=<0.2 % (Coulometric)
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
Chloride (Cl)=<50 ppm
Lead (Pb)=<0.1 ppm
Copper (Cu)=<0.1 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only