Tungsten hexacarbonyl, 97%
Tungsten hexacarbonyl, 97%
Tungsten hexacarbonyl, 97%
Thermo Scientific Chemicals

Tungsten hexacarbonyl, 97%

Tungsten hexacarbonyl, 99%, C6O6W, CAS Number-14040-11-0, 250g, 175.0 deg.C, Yellow to Beige, 237-880-2, 98.5% min., 06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51, 351.91, GHS Signal Word: Danger, GHS H Statement Toxic in contact with skin. Toxic if swallowed. Toxic if inhaled. | CAS: 14040-11-0 | C6O6W | 351.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 221040500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14040-11-0
IUPAC Nametungsten hexacarbonyl
Molecular FormulaC6O6W
InChI KeyFQNHWXHRAUXLFU-UHFFFAOYSA-N
SMILES[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only