Nalgene™ HDPE, Rectangular Carboy
Thermo Scientific™

Nalgene™ HDPE, Rectangular Carboy

For Use With: Convenient for dry materials such as solids and powders; Closure Material: Polypropylene; Color: White
Have Questions?
Número do catálogoCapacity (Metric)
2211-005020 L
Número do catálogo 2211-0050
Preço (BRL)
-
Capacity (Metric):
20 L

Easily fill, transfer and clean using the wide-mouth opening of Thermo Scientific™ Nalgene™ HDPE Rectangular Carboys without Spigots. Featuring a sturdy, space-saving design, these high density polyethylene carboys have convenient molded-in graduations to deliver in liters and gallons.

  • Graduated to contain in 1 gal. and 5 L increments for ease of us
  • Convenient for dry materials such as solids and powders
  • Leakproof†

Includes:

  • Polypropylene 100 mm screw closures

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for two minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Especificações
Capacity (English)5.3 gal.
Closure MaterialPolypropylene
Closure Size100-415 mm
Dimensions (L x W x H)12.5 x 8.63 x 15 in. (318 x 219 x 381 mm)
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
No. per Case4/Cs.
No. per Pack1 Ea.
Workflow StepUpstream process solution storage. Upstream Validation
AutoclavableNot Autoclavable
Capacity (Metric)20 L
ColorWhite
MaterialHDPE
Product TypeRectangular Carboy
SterilitySterile
Unit SizeCase of 4