Applied Biosystems™

O-Ring, 5/64ID x 13/64OD x 1/16 BUNA, 310

Catalog number:  221100

 Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a BUNA O-Ring for the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Product Size: 1 unit
Shipping Condition: Room Temperature
Type: O-rings

Contents & storage

Store at room temperature.

Documents

Manuals & protocols