Nalgene™ Polypropylene, Rectangular Carboy with Closure
Thermo Scientific™

Nalgene™ Polypropylene, Rectangular Carboy with Closure

For Use With: Store solutions and handle large volumes of powders and other solid samples; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)
2212-00209 L
2212-005020 L
2212-0050PK20 L
카탈로그 번호 2212-0020
제품 가격(KRW)
683,000
온라인 행사
Ends: 31-Dec-2026
758,000
할인액 75,000 (10%)
Case of 6
용량(미터법):
9 L
제품 가격(KRW)
683,000
온라인 행사
Ends: 31-Dec-2026
758,000
할인액 75,000 (10%)
Case of 6

Store solutions and handle large volumes of powders and other solid samples with Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure. Featuring a space-saving design, a wide mouth opening for filling and cleaning and a sturdy stainless-steel handle, these carboys are tough, translucent and autoclavable.

Features:

  • Wide-mouth opening for easy filling, transferring and cleaning of carboys
  • Molded in graduations in 1 gallon and 5 L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Autoclavable, graduated and leakproof†

Includes:

  • Polypropylene screw closure

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

사양
용량(영국식 단위)2.4 gal.
마개 재질Polypropylene
마개 크기100-415 mm
치수(길이 x 폭 x 높이)9 x 6 x 14 in. (220 x 153 x 360 mm)
용도(애플리케이션)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
케이스당 수6/Cs.
팩당 수1 Ea.
워크플로우 단계Upstream process solution storage. Upstream Validation
오토클레이브 가능Autoclavable
용량(미터법)9 L
색상White
물질Polypropylene
제품 유형Rectangular Carboy
멸균Non-sterile
Unit SizeCase of 6