Potassium thioacetate, 98%
Potassium thioacetate, 98%
Potassium thioacetate, 98%
Thermo Scientific Chemicals

Potassium thioacetate, 98%

CAS: 10387-40-3 | C2H3KOS | 114.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 221300250
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.60
Save 9.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.60
Save 9.95
Each
Chemical Identifiers
CAS10387-40-3
IUPAC Nameethanethioic S-acid potassium
Molecular FormulaC2H4KOS
InChI KeySDJHDRMYZQFJJO-UHFFFAOYSA-N
SMILES[K].CC(S)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Titration with HClO497.5 to 102.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only