Tungsten(VI) chloride, 99.9+%, (trace metal basis)
Tungsten(VI) chloride, 99.9+%, (trace metal basis)
Tungsten(VI) chloride, 99.9+%, (trace metal basis)
Thermo Scientific Chemicals

Tungsten(VI) chloride, 99.9+%, (trace metal basis)

CAS: 13283-01-7 | Cl6W | 396.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 221430100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13283-01-7
IUPAC Nametungsten hexachloride
Molecular FormulaCl6H6W
InChI KeyXHYHOBJDYHRWHO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.Cl.Cl.[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.10 %
Appearance (Color)Grey to dark blue to violet to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only