Phenetole, 98+%
Phenetole, 98+%
Phenetole, 98+%
Thermo Scientific Chemicals

Phenetole, 98+%

CAS: 103-73-1 | C8H10O | 122.17 g/mol
Quantity:
100 mL
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 221495000
Price (JPY)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-73-1
IUPAC Nameethoxybenzene
Molecular FormulaC8H10O
InChI KeyDLRJIFUOBPOJNS-UHFFFAOYSA-N
SMILESCCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5055 to 1.5085 (20°C, 589 nm)
GC>=98.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only