Phenol, 99.5%, extra pure, loose crystals, unstabilized
Phenol, 99.5%, extra pure, loose crystals, unstabilized
Phenol, 99.5%, extra pure, loose crystals, unstabilized
Thermo Scientific Chemicals

Phenol, 99.5%, extra pure, loose crystals, unstabilized

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Catalog NumberQuantity
2217500151.5 kg
Catalog number 221750015
Price (USD)
-
Quantity:
1.5 kg
Request bulk or custom format
Chemical Identifiers
CAS108-95-2
IUPAC Namephenol
Molecular FormulaC6H6O
InChI KeyISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Loose crystals or flakes
Residue after evaporation=<0.05 %
Appearance of solution(per Ph. Eur.) Passes test
Infrared spectrumConforms
Water=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only