2-Chloroethyl methyl ether, 98%
2-Chloroethyl methyl ether, 98%
2-Chloroethyl methyl ether, 98%
Thermo Scientific Chemicals

2-Chloroethyl methyl ether, 98%

CAS: 627-42-9 | C3H7ClO | 94.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 221770025
Price (USD)
1,286.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
1,286.00
Each
Chemical Identifiers
CAS627-42-9
IUPAC Name1-chloro-2-methoxyethane
Molecular FormulaC3H7ClO
InChI KeyXTIGGAHUZJWQMD-UHFFFAOYSA-N
SMILESCOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4070 to 1.4110 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only