Methyl 3-bromopropionate, 97%
Methyl 3-bromopropionate, 97%
Methyl 3-bromopropionate, 97%
Thermo Scientific Chemicals

Methyl 3-bromopropionate, 97%

CAS: 3395-91-3 | C4H7BrO2 | 167.00 g/mol
Quantity:
25 mL
50 mL
100 mL
Packaging:
Glass bottle
Catalog number 221791000
Price (JPY)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3395-91-3
IUPAC Namemethyl 3-bromopropanoate
Molecular FormulaC4H7BrO2
InChI KeyKQEVIFKPZOGBMZ-UHFFFAOYSA-N
SMILESCOC(=O)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4565 to 1.4595 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only