Benzoic acid, 99.5%, for analysis
Benzoic acid, 99.5%, for analysis
Benzoic acid, 99.5%, for analysis
Thermo Scientific Chemicals

Benzoic acid, 99.5%, for analysis

CAS: 65-85-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 221800050
Price (EUR)
528,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
528,00
Each
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point121°C to 123°C
Sulfated ash=<0.01 %
Iron (Fe)=<4 ppm
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only