Dionex™ Inuvion™ IC System
Dionex™ Inuvion™ IC System
Dionex™ Inuvion™ IC System
Thermo Scientific™
Dionex™ Inuvion™ IC System
Green features
Get consistent results and eliminate the need for chemical reagents with the Thermo Scientific™ Dionex™ Inuvion™ IC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
22185-60108Dionex Inuvion IC System with RFIC
22185-60104Dionex Inuvion IC System
Catalog number 22185-60108
Price (BRL)
-
Includes:
Dionex Inuvion IC System with RFIC
Make ion analysis simpler and more intuitive with the Thermo Scientific™ Dionex™ Inuvion™ IC system. Delivering consistently excellent results, the smart, function-driven design of the Dionex Inuvion IC system lets operators quickly and safely access everything on the instrument without hard-to-reach compartments, step stools, or extra tools. This system provides eluent degas and electrolytic suppression capabilities, with the option to add Reagent-Free IC (RFIC™) eluent generation for more consistent results and gradient separation capabilities.
  • The thermostatted high-performance conductivity detector permits measurements that are unaffected by temperature variation for improved reproducibility.
  • Includes advanced single-range digital output with operating range to 18,000 μS full scale, with autoranging to provide accurate detection of major and minor constituents in a single run. Single-range analog signal output is also standard.
  • Inert, nonmetallic, PEEK components throughout the system ensure compatibility with corrosive eluents and provide metal-contamination-free chromatography.
  • An electronically actuated six-port Rheodyne PEEK injection valve provides precise sampling.
  • Electrolytically regenerated or chemically regenerated suppression options offer application flexibility.
  • This system comes with the option to upgrade to RFIC eluent generation to broaden the scope of your applications.
Specifications
DescriptionDionex Inuvion IC System with RFIC
Green FeaturesACT Ecolabel (Silver)
IncludesDionex Inuvion IC System with RFIC
Max. Pressure35 MPa (5000 psi)
Detector TypeConductivity (CD)
Product TypeRFIC System
Unit SizeEach
Frequently asked questions (FAQs)
How does RFIC technology improve analytical consistency?
By eliminating manual eluent preparation, RFIC technology ensures consistent eluent strength and composition, enhancing retention time reproducibility, peak area reproducibility, and system sensitivity.
What is different about the Dionex Inuvion model with RFIC?
The RFIC (Reagent-Free Ion Chromatography) version of the Dionex Inuvion IC system includes automated eluent generation. This feature uses Thermo Scientific Dionex EGC Eluent Generator Cartridges to generate eluent. Users simply add deionized water. The system then precisely produces hydroxide, carbonate, bicarbonate, or methanesulfonic acid (MSA) eluents on demand. This simplifies operation, reduces contamination risks, and supports gradient separations.
What makes electrolytic suppression a better choice than chemical suppression?
Electrolytic suppression delivers more consistent and stable results by generating the suppressor reagent automatically, eliminating the need for manual chemical handling or replenishment. This reduces maintenance, improves long-term accuracy, and ensures more reliable performance across a wide range of applications.
What distinguishes the Dionex Inuvion IC system from the Dionex Inuvion Core version?
The Dionex Inuvion IC system adds electrolytic suppression and vacuum degassing of the eluent which reduces noise and improves consistency. It’s an ideal mid-level system for users seeking a higher level of automation and performance stability.
What accessories can I add to the Dionex Inuvion IC system?
We offer multiple options and peripherals to extend the capabilities of the Dionex Inuvion IC system. Available accessories include:
  • An additional 6-port or 10-port valve
  • Column heater
  • Digital gas pressure regulator
  • IC PEEK tubing with Viper fittings
  • An integrated regenerant pump
  • Seal wash pump
  • Eluent monitor (for 2 L or 4 L bottles)
You can easily swap these accessories in and out of the instrument in less than 5 minutes, allowing you to customize the system as your application needs change.