Dionex ™ Inuvion™ System Upgrades
Dionex ™ Inuvion™ System Upgrades
Dionex ™ Inuvion™ System Upgrades
Dionex ™ Inuvion™ System Upgrades
Thermo Scientific™
Dionex ™ Inuvion™ System Upgrades
Extend the capabilities and performance of your Thermo Scientific™ Dionex™ Inuvion™ system with a system upgrade.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
22185-62001Upgrade
22185-62002RFIC Upgrade
Catalog number 22185-62001
Price (EUR)
12.100,00
Each
Description:
Upgrade
Price (EUR)
12.100,00
Each
Extend the capabilities and performance of your Thermo Scientific™ Dionex™ Inuvion™ IC system with a system upgrade. Add electrolytic suppression and eluent degas to your Thermo Scientific™ Dionex™ Inuvion™ Core IC system to convert it to a full Dionex Inuvion IC system. You can also upgrade your Dionex Inuvion IC system by adding RFIC™ eluent generation to take advantage of the full benefits of reagent-free ion chromatography.
Benefits of a Dionex Inuvion Core IC system to Dionex Inuvion IC system upgrade
  • Electrolytic suppression
  • Eluent degas
Benefits of a Dionex Inuvion IC system to Dionex Inuvion IC system with RFIC upgrade
  • Electrolytic eluent generation with RFIC EG staging area and EG degasser
Note: This option is not available for the Dionex Inuvion Core IC system unless it has already been upgraded to the Dionex Inuvion IC system.
Specifications
DescriptionUpgrade
Product TypeUpgrade Kit
Unit SizeEach