1-Bromo-2-iodobenzene, 99%, stabilized
1-Bromo-2-iodobenzene, 99%, stabilized
1-Bromo-2-iodobenzene, 99%, stabilized
Thermo Scientific Chemicals

1-Bromo-2-iodobenzene, 99%, stabilized

CAS: 583-55-1 | C6H4BrI | 282.9 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 221950050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS583-55-1
IUPAC Name1-bromo-2-iodobenzene
Molecular FormulaC6H4BrI
InChI KeyOIRHKGBNGGSCGS-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.6620 to 1.6650 (20°C, 589 nm)
StabilizerCopper
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only