Sulfamic acid, 99%
Sulfamic acid, 99%
Sulfamic acid, 99%
Thermo Scientific Chemicals

Sulfamic acid, 99%

CAS: 5329-14-6 | H3NO3S | 97.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 222070025
Price (EUR)
238,65
Online exclusive
265,00
Save 26,35 (10%)
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
238,65
Online exclusive
265,00
Save 26,35 (10%)
Each
Chemical Identifiers
CAS5329-14-6
IUPAC Namesulfamic acid
Molecular FormulaH3NO3S
InChI KeyIIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILESNS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point200°C to 208°C
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only