1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy
1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy
1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy
1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy

CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 222085000
Price (USD)
104.65
Special offer
Online exclusive
Ends: 28-Dec-2026
123.00
Save 18.35
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
104.65
Special offer
Online exclusive
Ends: 28-Dec-2026
123.00
Save 18.35
Each
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 340 nm A: =<0.01
Appearance (Form)Liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.0 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only