Pierce™ Mannan Binding Protein Agarose

Catalog number: 22212B

Thermo Scientific™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5 mL column of the MBP Agarose.

Mannan binding protein (MBP), also known as mannose-binding lectin (MBL) is a mammalian lectin that is produced in the liver and occurs in serum. MBP consists of 18 identical subunits, each with a molecular mass of approximately 31kDa. The protein is capable of initiating carbohydrate-mediated complement activation.

Related Products
Pierce™ IgM Purification Kit
MBP Column Preparation Buffer for Pierce™ IgM Purification Kit
Elution Buffer for Pierce™ IgM Purification Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Affinity, Agarose Resin
Format: Bottle(s)
Immobilized Phase: Lectin
Product Line: Pierce™
Product Size: Custom

Contents & storage

Upon receipt store product at 4°C. Product is shipped at ambient temperature.

Documents

Manuals & protocols