Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Thermo Scientific Chemicals

Acetic anhydride, 99+%, for analysis

CAS: 108-24-7 | C4H6O3 | 102.089 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
檢視全部
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 222130010
價格 (TWD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS108-24-7
IUPAC Nameacetyl acetate
Molecular FormulaC4H6O3
InChI KeyWFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILESCC(=O)OC(C)=O
檢視更多
規格Specification Sheet規格表
Appearance (Form)Clear liquid
Chloride (Cl)=<5 ppm
Sulfate (SO4)=<5 ppm
Calcium (Ca)=<0.5 ppm
Copper (Cu)=<0.02 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent for analysis
RUO – Research Use Only