Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Acetic anhydride, 99+%, for analysis
Thermo Scientific Chemicals

Acetic anhydride, 99+%, for analysis

CAS: 108-24-7 | C4H6O3 | 102.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 222130100
Price (TWD)
-
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS108-24-7
IUPAC Nameacetyl acetate
Molecular FormulaC4H6O3
InChI KeyWFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILESCC(=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Chloride (Cl)=<5 ppm
Sulfate (SO4)=<5 ppm
Calcium (Ca)=<0.5 ppm
Copper (Cu)=<0.02 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent for analysis
RUO – Research Use Only