Acetic Acid, 99.8%, for analysis
Acetic Acid, 99.8%, for analysis
Acetic Acid, 99.8%, for analysis
Thermo Scientific Chemicals

Acetic Acid, 99.8%, for analysis

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
数量:
250 mL
1 L
2.5 L
5 L
20 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 222140025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS64-19-7
IUPAC Nameacetic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
さらに表示
Sulfate (SO4)=<1 ppm
Acetic anhydride (C4H6O3)=<0.01 %
Residue after evaporation=<0.001 %
Appearance (Form)Clear liquid
Titration with NaOH>=99.8 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only