Tetrahydrofuran, 99.5+%, for spectroscopy
Tetrahydrofuran, 99.5+%, for spectroscopy
Tetrahydrofuran, 99.5+%, for spectroscopy
Thermo Scientific Chemicals

Tetrahydrofuran, 99.5+%, for spectroscopy

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 222162500
Price (USD)
63.65
Special offer
Online exclusive
Ends: 28-Dec-2026
74.20
Save 10.55
Each
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 28-Dec-2026
74.20
Save 10.55
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 215 nm A: =<1
UVat 270 nm A: =<0.04
Residue after evaporation=<0.005 %
Alkalinity=<0.0006 meq/g
Acidity=<0.0002 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only