Dimethyl fumarate, 99%
Dimethyl fumarate, 99%
Dimethyl fumarate, 99%
Thermo Scientific Chemicals

Dimethyl fumarate, 99%

CAS: 624-49-7 | C6H8O4 | 144.13 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 222181000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS624-49-7
IUPAC Name1,4-dimethyl (2E)-but-2-enedioate
Molecular FormulaC6H8O4
InChI KeyLDCRTTXIJACKKU-ONEGZZNKSA-N
SMILESCOC(=O)\C=C\C(=O)OC
さらに表示
Appearance (Color)White to light beige to light brown
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Free acid=<1.5 % (as Fumaric acid)
Melting point100°C to 107°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only