Thermo Scientific Chemicals

Dimethyl fumarate, 99%, Thermo Scientific Chemicals

Catalog number: 222185000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl fumarate, 99%, Thermo Scientific Chemicals

Catalog number: 222185000
500 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
624-49-7
IUPAC Name
1,4-dimethyl (2E)-but-2-enedioate
Molecular Formula
C6H8O4
InChI Key
LDCRTTXIJACKKU-ONEGZZNKSA-N
SMILES
COC(=O)\C=C\C(=O)OC
Appearance (Color)
White to light beige to light brown
Melting point
100°C to 107°C
Appearance (Form)
Crystalline powder or crystals or flakes
Infrared spectrum
Conforms
Free acid
=<1.5 % (as Fumaric acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text