Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Thermo Scientific Chemicals

Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade

CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 222220250
Price (EUR)
276,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
276,00
Each
Chemical Identifiers
CAS6152-67-6
IUPAC Name4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular FormulaC12H11NNaO3S
InChI KeyVMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Off-white to beige to light grey
Infrared spectrumConforms
Sensitivity(as indicator) Passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only