Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Thermo Scientific Chemicals

Poly(sodium-p-styrenesulfonate), average M.W. 70.000

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 222270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS25704-18-1
Molecular Formula(C8H7NaO3S)n
SMILES[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*
MDL NumberMFCD00084449
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only