Chloromethanesulfonyl chloride, 95%
Chloromethanesulfonyl chloride, 95%
Chloromethanesulfonyl chloride, 95%
Thermo Scientific Chemicals

Chloromethanesulfonyl chloride, 95%

CAS: 3518-65-8 | CH2Cl2O2S | 148.99 g/mol
製品番号(カタログ番号)数量パッケージ
2224600505 gGlass bottle
製品番号(カタログ番号) 222460050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3518-65-8
IUPAC Namechloromethanesulfonyl chloride
Molecular FormulaCH2Cl2O2S
InChI KeyKQDDQXNVESLJNO-UHFFFAOYSA-N
SMILESClCS(Cl)(=O)=O
さらに表示
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only