Strontium nitrate, 99+%, for analysis
Strontium nitrate, 99+%, for analysis
Strontium nitrate, 99+%, for analysis
Thermo Scientific Chemicals

Strontium nitrate, 99+%, for analysis

Strontium nitrate, 99+%, N2O6Sr, CAS Number-10042-76-9, bdg873aqzl, strontium dinitrate, nitric acid, strontium salt 2:1, hsdb 787, nitric acid, strontium salt, nitrate de strontium french, strontium nitrate sr no3 2, strontium nitrate, strontium ii nitrate 1:2, unii-bdg873aqzl, 1kg | CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 222562500
Price (EUR)
39,30
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
39,30
Each
Chemical Identifiers
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH5 to 7 (5 % at 20°C)
Chloride (Cl)=<10 ppm
Copper (Cu)=<2 ppm
Potassium (K)=<50 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only