Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Thermo Scientific Chemicals

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf

CAS: 14985-18-3 | N2O7Zr | 231.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 222631000
Price (EUR)
72,65
Online Exclusive
80,20
Save 7,55 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
72,65
Online Exclusive
80,20
Save 7,55 (9%)
Each
Chemical Identifiers
CAS14985-18-3
IUPAC Name(nitrooxy)(oxo)zirconio nitrate
Molecular FormulaN2O7Zr
InChI KeyJWBWFTANBHYHBC-UHFFFAOYSA-N
SMILES[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Gravimetrycalculated on the actual degree of hydration
Gravimetrycalculated on the actual degree of hydration
Gravimetry34.9 to 37.0 % (ZrO2)
Hafnium (Hf)=<4 %
Gravimetry>=99.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only