Thermo Scientific Chemicals

Barium acetate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 222800025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Barium acetate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 222800025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222800025
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
543-80-6
IUPAC Name
barium(2+) diacetate
Molecular Formula
C4H6BaO4
InChI Key
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES
[Ba++].CC([O-])=O.CC([O-])=O
Nitrate (NO3)
=<20 ppm
Magnesium (Mg)
=<10 ppm
Calcium (Ca)
=<50 ppm
Appearance (Form)
Crystalline powder
pH
6.5 to 8.5 (5 % at 20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell