Sodium metabisulfite, for analysis
Sodium metabisulfite, for analysis
Sodium metabisulfite, for analysis
Thermo Scientific Chemicals

Sodium metabisulfite, for analysis

Reduction reagent | CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol
数量:
500 g
1 kg
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 223075000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7681-57-4
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular FormulaNa2O5S2
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
さらに表示
Titration Iodimetric>=58.5 % (SO2)
Iron (Fe)=<20 ppm
Appearance (Color)White
Insoluble matter=<0.005 %
Appearance (Form)Crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only