Orion™ 2230XP Silica Analyzer Reagents and Standards
Actual product may vary
Thermo Scientific™
Orion™ 2230XP Silica Analyzer Reagents and Standards
The reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2230REReagents and Standard
223002TS
also known as 223002
20 ppb Calibration Standard
223010TS
also known as 223010
100 ppb Calibration Standard
223030TS
also known as 223030
1000 ppb Calibration Standard
Catalog number 2230RE
Price (USD)
297.00
Online exclusive
330.00
Save 33.00
Each
Type:
Reagents and Standard
Price (USD)
297.00
Online exclusive
330.00
Save 33.00
Each
Ensure you have the reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer. Reagents kit contains all three reagents required for operation, and calibration standards available in individual bottles. For use with the Orion 2230XP Silica Analyzer.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionReagents 1,2 and 3
Temperature (Metric) Operating5°C to 45°C
TypeReagents and Standard
Unit SizeEach