Nickel(II) nitrate hexahydrate, 99%, for analysis
Nickel(II) nitrate hexahydrate, 99%, for analysis
Nickel(II) nitrate hexahydrate, 99%, for analysis
Nickel(II) nitrate hexahydrate, 99%, for analysis
Thermo Scientific Chemicals

Nickel(II) nitrate hexahydrate, 99%, for analysis

CAS: 13478-00-7 | H12N2NiO12 | 290.79 g/mol
数量:
500 g
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 223150025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13478-00-7
IUPAC Namenickel(2+) hexahydrate dinitrate
Molecular FormulaH12N2NiO12
InChI KeyAOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Titration Complexometric>=98.5 %
Cadmium (Cd)=<5 ppm
Iron (Fe)=<10 ppm
Chromium (Cr)=<5 ppm
Aluminium (Al)=<10 ppm
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For analysis
RUO – Research Use Only