Titanium(IV) n-butoxide, 98+%
Titanium(IV) n-butoxide, 98+%
Titanium(IV) n-butoxide, 98+%
Thermo Scientific Chemicals

Titanium(IV) n-butoxide, 98+%

CAS: 5593-70-4 | C16H36O4Ti | 340.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
2 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 223190020
Price (USD)
226.65
Special offer
Online exclusive
Ends: 28-Dec-2026
266.00
Save 39.35
Each
Quantity:
2 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
226.65
Special offer
Online exclusive
Ends: 28-Dec-2026
266.00
Save 39.35
Each
Chemical Identifiers
CAS5593-70-4
IUPAC Nametitanium(4+) tetrakis(butan-1-olate)
Molecular FormulaC16H36O4Ti
InChI KeyYHWCPXVTRSHPNY-UHFFFAOYSA-N
SMILES[Ti+4].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
Titanium (Ti)13.8 to 14.4 %
Viscosity50 to 90 cP (at 25°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only