Product Image
Gibco™

DMEM, low glucose, pyruvate, HEPES

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. With L-Glutamine, 1000 mg/L D-Glucose, Sodium Pyruvate, 25 mM HEPES.
Have Questions?
Catalog NumberQuantity
2232003010 x 500 mL
Catalog number 22320030
Price (MXN)
-
Quantity:
10 x 500 mL
Customize this product

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: Low Glucose, L-glutamine, Sodium Pyruvate, HEPES, Phenol Red

The complete formulation is available.

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.

DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Concentration1 X
Culture EnvironmentCO2
Glucose Concentration1000 mg/L
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality250 - 290 mOsm/kg
Product LineGibco
Product TypeDulbecco's Modified Eagle Medium (DMEM)
Quantity10 x 500 mL
Shelf Life12 Months
ClassificationAnimal Origin-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesLow Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
pH7 to 7.4
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.

Frequently asked questions (FAQs)

What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.