Sodium chlorate, 99+%, extra pure
Sodium chlorate, 99+%, extra pure
Sodium chlorate, 99+%, extra pure
Thermo Scientific Chemicals

Sodium chlorate, 99+%, extra pure

CAS: 7775-09-9 | ClNaO3 | 106.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 223222500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7775-09-9
IUPAC Namesodium chlorate
Molecular FormulaClNaO3
InChI KeyYZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES[Na+].[O-][Cl](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Insoluble matter=<0.005 % (in water)
Appearance (Color)White to light yellow
Sodium chloride (NaCl)=<1000 ppm
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only