Hydrobromic acid, for analysis, ca. 48% solution in water
Hydrobromic acid, for analysis, ca. 48% solution in water
Hydrobromic acid, for analysis, ca. 48% solution in water
Thermo Scientific Chemicals

Hydrobromic acid, for analysis, ca. 48% solution in water

CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 223325000
Price (USD)
99.70
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
99.70
Each
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<500 ppm
Phosphate (PO4)=<10 ppm
Arsenic (As)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only