Product Image
Gibco™

Medium 199, HEPES

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivationLeia mais
Have Questions?
Número do catálogoQuantity
22340087
conhecido também como 22340-087
10 x 500 mL
Número do catálogo 22340087
conhecido também como 22340-087
Preço (BRL)
-
Quantity:
10 x 500 mL
Customize this product

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Medium 199 is modified as follows:

With: L-glutamine, Phenol Red, HEPES

The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.

Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Especificações
Cell TypeMammalian Cells
Concentration1 X
Culture EnvironmentCO2
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 - 330 mOsm/kg
Product LineGibco
Product TypeMedium 199
Quantity10 x 500 mL
Shelf Life12 Months
ClassificationAnimal Origin
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesGlutamine, HEPES, Phenol Red, Sodium Bicarbonate
Without AdditivesNo Sodium Pyruvate
pH7.1 to 7.3
Unit SizeEach
Conteúdo e armazenamento
Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.