5-Chloro-1-pentyne, 98%
5-Chloro-1-pentyne, 98%
5-Chloro-1-pentyne, 98%
5-Chloro-1-pentyne, 98%
Acros Organics

5-Chloro-1-pentyne, 98%

CAS: 14267-92-6 | C5H7Cl | 102.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 223420050
also known as 22342
HSN 29032900
Price (INR)
5,255.00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,255.00
Each
Add to cart
Chemical Identifiers
CAS14267-92-6
IUPAC Name5-chloropent-1-yne
Molecular FormulaC5H7Cl
InChI KeyUXFIKVWAAMKFQE-UHFFFAOYSA-N
SMILESClCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Refractive index1.4430 to 1.4450 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only