1-Decyne, 98%
1-Decyne, 98%
1-Decyne, 98%
Thermo Scientific Chemicals

1-Decyne, 98%

CAS: 764-93-2 | C10H18 | 138.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 223430250
Price (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS764-93-2
IUPAC Namedec-1-yne
Molecular FormulaC10H18
InChI KeyILLHQJIJCRNRCJ-UHFFFAOYSA-N
SMILESCCCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only