3,3-Dimethyl-1-butyne, 98%
3,3-Dimethyl-1-butyne, 98%
3,3-Dimethyl-1-butyne, 98%
Thermo Scientific Chemicals

3,3-Dimethyl-1-butyne, 98%

CAS: 917-92-0 | C6H10 | 82.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 223440050
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.80
Save 10.15
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.80
Save 10.15
Each
Chemical Identifiers
CAS917-92-0
IUPAC Name3,3-dimethylbut-1-yne
Molecular FormulaC6H10
InChI KeyPPWNCLVNXGCGAF-UHFFFAOYSA-N
SMILESCC(C)(C)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3730 to 1.3750 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only