Sulfuro de cinc, 99,99 %, (base de trazas metálicas), < 10 micrómetros, Thermo Scientific Chemicals
Sulfuro de cinc, 99,99 %, (base de trazas metálicas), < 10 micrómetros, Thermo Scientific Chemicals
Sulfuro de cinc, 99,99 %, (base de trazas metálicas), < 10 micrómetros, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfuro de cinc, 99,99 %, (base de trazas metálicas), < 10 micrómetros, Thermo Scientific Chemicals

CAS: 1314-98-3 | SZn | 97.44 g/mol
Have Questions?
Número de catálogoCantidadEnvase
22365025025 gFrasco de vidrio
Número de catálogo 223650250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1314-98-3
IUPAC Namezinc(2+) sulfanediide
Molecular FormulaSZn
InChI KeyDRDVZXDWVBGGMH-UHFFFAOYSA-N
SMILES[S--].[Zn++]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Total trace metal impurities=<0.01 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Para análisis de ultratrazas
RUO – Research Use Only