Tin(IV) oxide, 99.9%, (trace metal basis)
Tin(IV) oxide, 99.9%, (trace metal basis)
Tin(IV) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Tin(IV) oxide, 99.9%, (trace metal basis)

CAS: 18282-10-5 | O2Sn | 150.71 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 223685000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18282-10-5
IUPAC Namestannanedione
Molecular FormulaO2Sn
InChI KeyXOLBLPGZBRYERU-UHFFFAOYSA-N
SMILESO=[Sn]=O
さらに表示
Appearance (Color)White to light grey
Total trace metal impurities=<0.1 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only