Adjustable Reinforced Shelf with Channels
Adjustable Reinforced Shelf with Channels
Adjustable Reinforced Shelf with Channels
Adjustable Reinforced Shelf with Channels
Adjustable Reinforced Shelf with Channels
Thermo Scientific™
Adjustable Reinforced Shelf with Channels
Material: Stainless Steel; For Use With: Large capacity reach-in CO2 Incubator; ; Environmental chambers 3948, 3907 and 3962
Have Questions?
Número do catálogoType
224161Shelf
Número do catálogo 224161
Preço (BRL)
-
Type:
Shelf
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
Especificações
DescriptionReinforced Stainless Steel Shelf and Channels (max. load 68kg)
For Use With (Equipment)Large Capacity Reach-In CO2 Incubator
LightingLED
TypeShelf
MaterialStainless Steel
Unit SizeEach