2,6-Di-tert-butyl-4-methylpyridine, 98%
2,6-Di-tert-butyl-4-methylpyridine, 98%
2,6-Di-tert-butyl-4-methylpyridine, 98%
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylpyridine, 98%

CAS: 38222-83-2 | C14H23N | 205.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 224520050
Price (USD)
127.65
Special offer
Online exclusive
Ends: 28-Dec-2026
150.00
Save 22.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
127.65
Special offer
Online exclusive
Ends: 28-Dec-2026
150.00
Save 22.35
Each
Chemical Identifiers
CAS38222-83-2
IUPAC Name2,6-di-tert-butyl-4-methylpyridine
Molecular FormulaC14H23N
InChI KeyHVHZEKKZMFRULH-UHFFFAOYSA-N
SMILESCC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange to light brown
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4760 to 1.4780 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only