Nalgene™ Round Polycarbonate Clearboy™ Carboy with Closure
Nalgene™ Round Polycarbonate Clearboy™ Carboy with Closure
Thermo Scientific™
Nalgene™ Round Polycarbonate Clearboy™ Carboy with Closure
For Use With: Large-volume media and culture preparation, particularly when visual inspection is required.; Autoclavable; Closure Material: Polypropylene; Color: Clear
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2251-0050PK20 L
2251-002010 L
2251-0020PK10 L
2251-005020 L
Catalog number 2251-0050PK
Price (MXN)
7,619.39
Each
Add to cart
Capacity (Metric):
20 L
Price (MXN)
7,619.39
Each
Add to cart
Conduct large-volume media and culture preparation with these Nalgene™ Round Polycarbonate Clearboy™ Carboys, particularly when visual inspection is required. This polycarbonate Clearboy is durable, lighter and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.
Features:
  • Available in 10 and 20 L sizes for a variety of biopharmaceutical applications
  • Graduated in liters and gallons
  • Molded of transparent, durable polycarbonate
  • Customization options available for fluid transfer applications
  • Autoclavable, graduated, leakproof†
Includes:
  • Polypropylene closure and TPE gasket
Ordering Information:
Additional closure options include Nalgene Quick Filling/Venting Closures (Cat. No. 2158), Nalgene Filling/Venting Closures (Cat. No. 2162) Nalgene Top Works™ Fluid Transfer Systems (Cat. No. 2135) and Nalgene Validation Bottles (Cat. No. DS2127). Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149).
Note:
Completely disengage closure threads and disassemble spigot before autoclaving
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (English)5.3 gal.
Closure MaterialPolypropylene
Closure Size83B
Diameter (Metric) Outer287 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Height (Metric) With Closure536 mm
No. per Case4/Cs.
No. per Pack1 Ea.
ShapeRound
Volume (English) Brim6 gal.
Volume (Metric) Brim24 L
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage
AutoclavableAutoclavable
Capacity (Metric)20 L
ColorClear
MaterialPolycarbonate
Product TypeCarboy
SterilityNon-sterile
Unit SizeEach