1-Bromo-4-tert-butylbenzene, 97%
1-Bromo-4-tert-butylbenzene, 97%
1-Bromo-4-tert-butylbenzene, 97%
Thermo Scientific Chemicals

1-Bromo-4-tert-butylbenzene, 97%

CAS: 3972-65-4 | C10H13Br | 213.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 225191000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3972-65-4
IUPAC Name1-bromo-4-tert-butylbenzene
Molecular FormulaC10H13Br
InChI KeyXHCAGOVGSDHHNP-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.5315 to 1.5345 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only