N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
Thermo Scientific Chemicals

N,N-Dimethyloctylamine, 97%

CAS: 7378-99-6 | C10H23N | 157.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 225290025
Price (EUR)
2.345,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
2.345,00
Each
Chemical Identifiers
CAS7378-99-6
IUPAC Namedimethyl(octyl)amine
Molecular FormulaC10H23N
InChI KeyUQKAOOAFEFCDGT-UHFFFAOYSA-N
SMILESCCCCCCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<30 APHA
Titration with HClO4>=96.0 %
Appearance (Form)Clear to slightly hazy liquid
Infrared spectrumConforms
Refractive index1.4233 to 1.4253 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only