2-Aminoisobutyric acid, 98+%
2-Aminoisobutyric acid, 98+%
2-Aminoisobutyric acid, 98+%
Thermo Scientific Chemicals

2-Aminoisobutyric acid, 98+%

CAS: 62-57-7 | C4H9NO2 | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 225760250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS62-57-7
IUPAC Name2-amino-2-methylpropanoic acid
Molecular FormulaC4H9NO2
InChI KeyFUOOLUPWFVMBKG-UHFFFAOYSA-N
SMILESCC(C)(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=98.0 %
Infrared spectrumConforms
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only